Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ Fast Flow IEX Columns

Catalog No. 45002539 Shop All Cytiva Products
Click to view available options
Product Type:
HiScreen Q Sepharose FF Column
HiScreen SP Sepharose FF clumn
Ligand Type:
Quaternary Amine
Sulphopropyl
pH Range:
2 to 12
4 to 13

Catalog No. 45-002-539

Supplier: Cytiva 28950510

Only null left
Add to Cart
Add to Cart

Designed for method optimization and parameter screening

  • Ready-to-use column for convenient process development optimization and parameter screening
  • Highly cross-linked agarose matrix with average bead size of 90μm
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • 20cm bed height is acheived when it is connected in series
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity

  • Bed dimension: 7.7 × 100mm
  • Matrix: 6% cross-linked agarose beads
  • Average particle size: 90μm
  • Max. flow velocity: 450cm/hr.
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm

HiScreen Q FF

  • Media:
  • pH stability working range: 2 to 12
  • pH stability cleaning: 2 to 14
  • Ion exchanger type: strong anion exchanger
HiScreen SP FF
  • Media:
  • pH stability working range: 3 to 14
  • pH stability cleaning: 4 to 13
  • Ion exchanger type: strong cation exchanger

TRUSTED_SUSTAINABILITY

Specifications

Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Product Type HiScreen Q Sepharose FF Column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <450 cm/hr.
Material Polypropylene
Ligand Type Quaternary Amine
Max. Pressure 1.5 bar
pH Range 2 to 12
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Optimization and parameter screening, ion exchange chromato graphy
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.