Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H2AC Antibody, Novus Biologicals™
SDP

Catalog No. NBP145619 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP145619 0.1 mg
NB018571 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP145619 Supplier Novus Biologicals Supplier No. NBP145619

Rabbit Polyclonal Antibody

HIST1H2AC Polyclonal specifically detects HIST1H2AC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.

Specifications

Antigen HIST1H2AC
Applications Western Blot, Immunohistochemistry, Immunoprecipitation
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation
Formulation PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin 300
Gene Accession No. Q93077
Gene Alias dJ221C16.4, H2A/l, H2AFLmember L, histone 1, H2ac, histone cluster 1, H2ac, histone H2A type 1-C, Histone H2A/l, histone H2AC, MGC99519
Gene Symbols HIST1H2AC
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the N-terminus of human histone H2A.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 8334
Test Specificity It does not cross-react with other histones.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.