Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2AC Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP1456190.05MG
Description
HIST1H2AC Polyclonal specifically detects HIST1H2AC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
HIST1H2AC | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin | |
dJ221C16.4, H2A/l, H2AFLmember L, histone 1, H2ac, histone cluster 1, H2ac, histone H2A type 1-C, Histone H2A/l, histone H2AC, MGC99519 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
It does not cross-react with other histones. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/mL | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Q93077 | |
HIST1H2AC | |
Synthetic peptide corresponding to the N-terminus of human histone H2A. | |
0.05 mg | |
Cancer | |
8334 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction