Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST1H2AC Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$627.00
Specifications
Antigen | HIST1H2AC |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot, Immunohistochemistry, Immunoprecipitation |
Applications | Western Blot |
Classification | Polyclonal |
Description
HIST1H2AC Polyclonal specifically detects HIST1H2AC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
HIST1H2AC | |
Western Blot, Immunohistochemistry, Immunoprecipitation | |
Polyclonal | |
Rabbit | |
Cancer | |
PBS (pH 7.2), 5 mM EDTA, 0.5% BSA, 30% Glycerol with 0.03% Proclin | |
dJ221C16.4, H2A/l, H2AFLmember L, histone 1, H2ac, histone cluster 1, H2ac, histone H2A type 1-C, Histone H2A/l, histone H2AC, MGC99519 | |
HIST1H2AC | |
IgG | |
Affinity Purified | |
It does not cross-react with other histones. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
Q93077 | |
8334 | |
Synthetic peptide corresponding to the N-terminus of human histone H2A. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title