Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H2BK Rabbit anti-Human, Clone: SN72-09, Novus Biologicals™
SDP

Catalog No. NBP267682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267682 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267682 Supplier Novus Biologicals Supplier No. NBP267682

Rabbit Monoclonal Antibody

HIST1H2BK Monoclonal specifically detects HIST1H2BK in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HIST1H2BK
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SN72-09
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry 1:50-1:100, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:100-1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:100-1:500
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias H2B histone family, member T, H2B K, H2B/S, H2BFAiii, H2BFT, H2bk, HIRA-interacting protein 1, HIRIP1, histone cluster 1, H2bk, histone family member, histone H2B type 1-K, MGC131989
Gene Symbols HIST1H2BK
Host Species Rabbit
Immunogen Synthetic peptide within human HIST1H2BK aa 1-49/126. (SwissProt: O60814 Human; SwissProt: P06899 Human; SwissProt: P23527 Human; SwissProt: P33778 Human; SwissProt: P57053 Human; SwissProt: P58876 Human; SwissProt: P62807 Human; SwissProt: Q16778 Human; SwissProt: Q5QNW6 Human; SwissProt: Q8N257 Human; SwissProt: Q93079 Human; SwissProt: Q99877 Human; SwissProt: Q99879 Human; SwissProt: Q99880 Human; Entrez Gene: 64647 Mouse; Entrez Gene: Q00729 Rat)
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 85236
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.