Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A Antibody (CL4968), Novus Biologicals™
SDP

Catalog No. NBP261643 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP261643 100 μL
NB393086 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP261643 Supplier Novus Biologicals Supplier No. NBP261643
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIST1H3A Monoclonal specifically detects HIST1H3A in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Histone H3T
Applications Immunohistochemistry
Classification Monoclonal
Clone CL4968
Conjugate Unconjugated
Dilution Immunohistochemistry 1:200 - 1:500
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias H3.4, H3/g, H3FT, H3t, HIST3H3, histone cluster 3, H3
Gene Symbols HIST1H3A
Host Species Mouse
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: PKDIQLARRIRGERA
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 8290
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.