Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST1H3A, p Ser10 Antibody, Novus Biologicals™
SDP

Catalog No. NB10097852 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10097852 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10097852 Supplier Novus Biologicals Supplier No. NB10097852
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HIST1H3A Polyclonal specifically detects HIST1H3A in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HIST1H3A (p Ser10)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:10000, Flow Cytometry 0.4 ug, Immunohistochemistry 1:2000 - 1:10000, Immunocytochemistry/Immunofluorescence 1:2000 - 1:10000, Immunohistochemistry-Paraffin 1:2000 - 1:10000, Immunocytochemistry
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. NP_003520.1
Gene Alias H3 histone family, member A, H3/A, H3FAHIST1H3E, H3FB, H3FC, H3FD, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3a, histone cluster 1, H3a, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Gene Symbols HIST1H3A
Host Species Rabbit
Immunogen Immunogen for A301-844A wthis antibody a synthetic phosphorylated peptide, which represented a portion of human Histone H3 surrounding serine 10 according to the numbering given in entry NP_003520.1 (GeneID 8350).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Protein Kinase, Stem Cell Markers, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8350
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Rat, D. melanogaster, Zebrafish, X. laevis, X. tropicalis, Chicken, Bovine, Rabbit, Pig, Orangutan, Staghorn coral_126732, Darling's disease fungus_5037, Mouse-ear cress, Yeast_284811, and Yellow koji mold_510516.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.