Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIST2H3A Antibody (6D3B9) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP252483 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP252483 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP252483 Supplier Novus Biologicals Supplier No. NBP252483
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HIST2H3A Monoclonal specifically detects HIST2H3A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.

Specifications

Antigen HIST2H3A
Applications Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry
Classification Monoclonal
Clone 6D3B9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready
Formulation PBS with 0.05% Sodium Azide
Gene Alias H3/n, H3/o, H3F2, H3FM, HIST2H3C, HIST2H3D, histone 2, H3a, histone cluster 2, H3a, histone H3.2, Histone H3/m, Histone H3/o
Gene Symbols HIST2H3A
Host Species Mouse
Immunogen Synthesized peptide fragment of human HH3 (AA: 121-136) expressed in E. Coli.
Molecular Weight of Antigen 15.4 kDa
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 333932
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.