Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HIST2H3A Antibody (6D3B9) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$560.50
Specifications
Antigen | HIST2H3A |
---|---|
Clone | 6D3B9 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry |
Description
HIST2H3A Monoclonal specifically detects HIST2H3A in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
HIST2H3A | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, CyTOF, Immunocytochemistry | |
Unconjugated | |
Mouse | |
Human, Mouse, Rat | |
H3/n, H3/o, H3F2, H3FM, HIST2H3C, HIST2H3D, histone 2, H3a, histone cluster 2, H3a, histone H3.2, Histone H3/m, Histone H3/o | |
HIST2H3A | |
IgG1 | |
Protein A or G purified |
6D3B9 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
PBS with 0.05% Sodium Azide | |
333932 | |
Synthesized peptide fragment of human HH3 (AA: 121-136) expressed in E. Coli. | |
Primary | |
15.4 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title