Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine Antibody, Novus Biologicals™
SDP

Catalog No. NB036604 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036604 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036604 Supplier Novus Biologicals Supplier No. NBP289013

Rabbit Polyclonal Antibody

Histamine Polyclonal specifically detects Histamine in Non-species specific samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Histamine
Applications ELISA, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Neat rabbit serum.
Gene Alias 2 (4 imidazolyl)ethylamine, 4 (2 aminoethyl) 1 3 diazole
Host Species Rabbit
Immunogen Histamine conjugated to KLH.
Purification Method Unpurified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.