Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine H3R Antibody (1008944) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB402287 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402287 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402287 Supplier Novus Biologicals Supplier No. FAB10200I
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Histamine H3R Monoclonal specifically detects Histamine H3R in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Histamine H3R
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 1008944
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3
Host Species Mouse
Immunogen Synthetic peptide containing human Histamine H3 R, Accession # Q9Y5N1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 11255
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.