Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine H3R Antibody (2420B) [Allophycocyanin], Novus Biologicals™
SDP

Catalog No. NB402309 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB402309 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB402309 Supplier Novus Biologicals Supplier No. FAB102001A
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histamine H3R Monoclonal antibody specifically detects Histamine H3R in Human, Mouse, Rat samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen Histamine H3R
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 2420B
Conjugate APC
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3
Host Species Rabbit
Immunogen Synthetic peptide containing Human/Mouse/Rat Histamine H3 R, Accession # Q9Y5N1
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 11255
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.