Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histamine H3R Antibody (2420B), HRP, Novus Biologicals™
SDP

Catalog No. NB340168 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340168 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340168 Supplier Novus Biologicals Supplier No. FAB102001H
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histamine H3R Monoclonal specifically detects Histamine H3R in Human, Mouse, Rat samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Histamine H3R
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 2420B
Conjugate HRP
Dilution Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3
Host Species Rabbit
Immunogen Synthetic peptide containing Human/Mouse/Rat Histamine H3 R, Accession # Q9Y5N1
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 11255
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.