Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H3R Rabbit anti-Human, Mouse, Rat, Clone: 7H8E6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP316202100UL
This item is not returnable.
View return policy
Description
Histamine H3R Monoclonal antibody specifically detects Histamine H3R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Histamine H3R | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
7H8E6 | |
Western Blot, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:500 | |
GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Histamine H3R (Q9Y5N1). | |
100 μg | |
GPCR, Immunology, Innate Immunity | |
11255 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction