Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histamine H3R Rabbit anti-Human, Mouse, Rat, Clone: 7H8E6, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Histamine H3R |
---|---|
Clone | 7H8E6 |
Dilution | Western Blot, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:500 |
Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Histamine H3R Monoclonal antibody specifically detects Histamine H3R in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
Histamine H3R | |
Western Blot, Immunohistochemistry 1:200 - 1:500, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Histamine H3R (Q9Y5N1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
7H8E6 | |
Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
GPCR, Immunology, Innate Immunity | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
11255 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title