Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histo-blood group ABO system transferase Antibody (SR1686), Novus Biologicals™
SDP

Catalog No. NB306088 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB306088 100 μL
NB306087 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB306088 Supplier Novus Biologicals Supplier No. NBP322267100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histo-blood group ABO system transferase Monoclonal antibody specifically detects Histo-blood group ABO system transferase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Histo-blood group ABO system transferase
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1686
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Host Species Rabbit
Immunogen Synthetic peptide from N-terminal
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.