Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histo-blood group ABO system transferase Antibody (SR1686), Novus Biologicals™

Rabbit Monoclonal Antibody
$282.00 - $393.00
Specifications
Antigen | Histo-blood group ABO system transferase |
---|---|
Clone | SR1686 |
Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
Applications | Western Blot, Immunoprecipitation |
Classification | Monoclonal |
Description
Histo-blood group ABO system transferase Monoclonal antibody specifically detects Histo-blood group ABO system transferase in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
Histo-blood group ABO system transferase | |
Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
IgG | |
Affinity purified |
SR1686 | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Synthetic peptide from N-terminal | |
Primary | |
Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title