Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Acetyltransferase Inhibitor IV, CPTH2, PROMO, Thermo Scientific Chemicals

Catalog No. J65939MAPM
Click to view available options
Quantity:
10 mg

A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 357649-93-5
Molecular Formula C14H14ClN3S
Synonym HAT Inhibitor IV; Gcn5p Inhibitor

Specifications

Quantity 10 mg
Content And Storage 2°C to 8°C
For Use With (Application) A histone acetyltransferase (HAT) inhibitor modulating the Gcn5 network where HATs act as transcriptional coactivators
Sensitivity Light Sensitive
Solubility Soluble in DMSO
Product Type Signal Transduction Reagent
Form Solid

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.