Learn More
Description
Specifications
Specifications
| Antigen | Histone Deacetylase 2/HDAC2 |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, ChIP Assay, KnockDown |
| Classification | Monoclonal |
| Clone | C.15200201 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1 ug, Chromatin Immunoprecipitation Sequencing 1ug/ChIP, Knockdown Validated |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1 |
| Gene Symbols | HDAC2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
