Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase 2/HDAC2 Antibody, DyLight 680, Novus Biologicals™
SDP

Catalog No. NB120044 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120044 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120044 Supplier Novus Biologicals Supplier No. NBP144095FR
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone Deacetylase 2/HDAC2 Polyclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Histone Deacetylase 2/HDAC2
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1
Host Species Rabbit
Immunogen Histone Deacetylase 2/Histone Deacetylase 2/HDAC2 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to the near C-terminal region of human Histone Deacetylase 2/HDAC2. (Uniprot: Q92769)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3066
Target Species Human, Mouse, Rat, Primate
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.