Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase 2/HDAC2 Antibody (OTI7E10), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP270881UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP270881UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP270881UV Supplier Novus Biologicals Supplier No. NBP270881UV

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Histone Deacetylase 2/HDAC2 Monoclonal specifically detects Histone Deacetylase 2/HDAC2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Histone Deacetylase 2/HDAC2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7E10
Conjugate DyLight 350
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.5.1.98, HD2, histone deacetylase 2, RPD3, transcriptional regulator homolog RPD3, YAF1, YY1-associated factor 1
Gene Symbols HDAC2
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 415-488 of human Histone Deacetylase 2/HDAC2 (NP_001518) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Breast Cancer, Cancer, Chromatin Research, Epigenetics, Histone Deacetylases, Phospho Specific, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 3066.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.