Learn More
Description
Specifications
Specifications
| Antigen | Histone Deacetylase 4/HDAC4 |
| Applications | Chemotaxis, ChIP Assay, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5-2 ug/ml, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Caretti et al (2003); Imbriano (2005)), Immunoprecipitation assay dependent, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000 |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Accession No. | NP_848510 |
| Gene Alias | HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 |
| Gene Symbols | HDAC4 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
