Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase 4/HDAC4 Antibody, Novus Biologicals™
SDP

Catalog No. NB10056341T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056341T 0.025 mg
NB10056341 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056341T Supplier Novus Biologicals Supplier No. NB10056341SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 4 publications

Histone Deacetylase 4/HDAC4 Polyclonal specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse samples. It is validated for Chemotaxis, ChIP Assay, Immunoprecipitation, Western Blot.

Specifications

Antigen Histone Deacetylase 4/HDAC4
Applications Chemotaxis, ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.5-2 ug/ml, Chromatin Immunoprecipitation 1:20-1:1000. Use reported in scientific literature (Caretti et al (2003); Imbriano (2005)), Immunoprecipitation assay dependent, Chromatin Immunoprecipitation (ChIP) 1:20-1:1000
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. NP_848510
Gene Alias HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3
Gene Symbols HDAC4
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acids 194-209 of human HDAC4.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics, Histone Deacetylases
Primary or Secondary Primary
Gene ID (Entrez) 9759
Test Specificity A 140 kDa band is observed.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.