Learn More
Description
Specifications
Specifications
| Antigen | Histone Deacetylase 4/HDAC4 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Dilution | Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | PBS with No Preservative |
| Gene Alias | HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 |
| Gene Symbols | HDAC4 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide made to a C-terminal portion of the human HDAC4 protein (between residues 1000-1084). [UniProt P56524] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
