Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 4/HDAC4, p Ser632 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2546360.05MG
Description
Histone Deacetylase 4/HDAC4 Polyclonal specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence.Specifications
Histone Deacetylase 4/HDAC4 | |
Polyclonal | |
Unconjugated | |
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide | |
HDAC4 | |
HDAC4 (Ab-632) antibody was raised against a peptide sequence around aa. 630∼634 (A-Q-S-S-P) derived from Human HDAC4. | |
Primary | |
Human, Mouse, Rat | |
IgG |
Western Blot, Immunofluorescence | |
1 mg/mL | |
Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 | |
HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
Rabbit | |
0.05 mg | |
9759 | |
Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction