Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 4/HDAC4, p Ser632 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
$196.10 - $426.00
Specifications
Antigen | Histone Deacetylase 4/HDAC4 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25463605
![]() |
Novus Biologicals
NBP2546360.05MG |
0.05 mg |
Each for $426.00
|
|
|||||
NB413824
![]() |
Novus Biologicals
NBP2546360.025MG |
0.025 mg |
Each for $196.10
|
|
|||||
Description
Histone Deacetylase 4/HDAC4 Polyclonal specifically detects Histone Deacetylase 4/HDAC4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence.Specifications
Histone Deacetylase 4/HDAC4 | |
Western Blot 1:500-1:1000, Immunofluorescence 1:100-1:200 | |
Polyclonal | |
Rabbit | |
HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
HDAC4 | |
IgG | |
Store at -20°C. Avoid freeze/thaw cycles. |
1 mg/mL | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Human, Mouse, Rat | |
9759 | |
HDAC4 (Ab-632) antibody was raised against a peptide sequence around aa. 630∼634 (A-Q-S-S-P) derived from Human HDAC4. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title