Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase 4/HDAC4 Rabbit anti-Mouse, Rat, Clone: 9A6E2, Novus Biologicals™
SDP

Catalog No. NB166023 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166023 20 μg
NB166022 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB166023 Supplier Novus Biologicals Supplier No. NBP31546320UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone Deacetylase 4/HDAC4 Monoclonal antibody specifically detects Histone Deacetylase 4/HDAC4 in Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Histone Deacetylase 4/HDAC4
Applications Western Blot
Classification Monoclonal
Clone 9A6E2
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Histone Deacetylase 4/HDAC4 (P56524).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 9759
Target Species Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.