Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone Deacetylase 4/HDAC4 Rabbit anti-Mouse, Rat, Clone: 9A6E2, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $458.50
Specifications
Antigen | Histone Deacetylase 4/HDAC4 |
---|---|
Clone | 9A6E2 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Histone Deacetylase 4/HDAC4 Monoclonal antibody specifically detects Histone Deacetylase 4/HDAC4 in Mouse, Rat samples. It is validated for Western BlotSpecifications
Histone Deacetylase 4/HDAC4 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Breast Cancer, Cancer, Cell Cycle and Replication, Chromatin Research, Epigenetics | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
9759 | |
IgG | |
Affinity purified |
9A6E2 | |
Monoclonal | |
Purified | |
RUO | |
Mouse, Rat | |
HA6116, HD4EC 3.5.1.98, HDAC-A, HDACABDMR, histone deacetylase 4, histone deacetylase A, KIAA0288AHO3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Histone Deacetylase 4/HDAC4 (P56524). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title