Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone Deacetylase 8/HDAC8 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB317966 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB317966 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB317966 Supplier Novus Biologicals Supplier No. NBP298969MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone Deacetylase 8/HDAC8 Polyclonal antibody specifically detects Histone Deacetylase 8/HDAC8 in Human samples. It is validated for Western Blot

Specifications

Antigen Histone Deacetylase 8/HDAC8
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 3.5.1.98, HD8, HDACL1, histone deacetylase 8, histone deacetylase-like 1, RPD3
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Histone Deacetylase 8/HDAC8.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Chromatin Research, Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 55869
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.