Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1.3 Rabbit anti-Human, Mouse, Rat, Clone: 3K5C10, Novus Biologicals™
SDP

Catalog No. NB168933 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB168933 100 μg
NB168934 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB168933 Supplier Novus Biologicals Supplier No. NBP316927100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H1.3 Monoclonal antibody specifically detects Histone H1.3 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen Histone H1.3
Applications Western Blot
Classification Monoclonal
Clone 3K5C10
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias H1 histone family, member 3, H1.3, H1F3H1D, histone 1, H1d, histone cluster 1, H1d, histone H1.3, Histone H1c, MGC138176
Host Species Rabbit
Immunogen A synthetic phosphorylated peptide around T17 of human Histone H1.3 (P16402).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3007
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.