Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H1.3 Rabbit anti-Human, Mouse, Rat, Clone: 3K5C10, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $516.00
Specifications
Antigen | Histone H1.3 |
---|---|
Clone | 3K5C10 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Histone H1.3 Monoclonal antibody specifically detects Histone H1.3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Histone H1.3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Epigenetics | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3007 | |
IgG | |
Affinity purified |
3K5C10 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
H1 histone family, member 3, H1.3, H1F3H1D, histone 1, H1d, histone cluster 1, H1d, histone H1.3, Histone H1c, MGC138176 | |
A synthetic phosphorylated peptide around T17 of human Histone H1.3 (P16402). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title