Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1 Antibody, Novus Biologicals™
SDP

Catalog No. NB017817 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.125 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017817 0.025 mg
NB100748 0.125 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017817 Supplier Novus Biologicals Supplier No. NB1007480.025MG
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Histone H1 Polyclonal specifically detects Histone H1 in Human, Mammalia, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen Histone H1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.3mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:300, Immunohistochemistry 1:100, Immunoprecipitation 1:20, Immunohistochemistry-Paraffin 1:100
Formulation PBS (pH 7.2) with No Preservative
Gene Alias H1 histone family, member 1, H1.1, H1A, H1F1HIST1, histone 1, H1a, histone cluster 1, H1a, histone H1.1, MGC126642, MGC138345
Gene Symbols HIST1H1A
Host Species Sheep
Immunogen Calf thymus intact Histone H1 (MW 23 kDa) complexed to rRNA.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 3024
Test Specificity This specifically binds to the Histone H1 protein found in nucleosomes of all eukaryotic cells. Differences between H1 subfractions of one species are greater than the differences across species. No cross reactivity was observed with other Histone core proteins H2A, H2B, H3 or H4.
Target Species Human, Mouse, Mammalia, Mouse, Rat
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.