Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1, Mouse anti-Drosophila, Clone: 1G6, Millipore Sigma™

Catalog No. p-7162910 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE188525 25 μg
MABE1885100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABE188525 Supplier MilliporeSigma Supplier No. MABE188525UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Histone H1, clone 1G6 Antibody, Cat. No. MABE1885, is a highly specific mouse monoclonal antibody that targets Drosophila Histone H1 and has been tested for use in Chromatin Immunoprecipitation (ChIP), ChIP-seq, Immunoprecipitation, and Western Blotting.

Histone H1 (UniProt: P02255; also known as H1) is encoded by the His 1 gene (Gene ID: 318854) in Drosophila species. Histone H1 is a linker histone that is involved in the organization and maintenance of chromatin higher order structure and plays an active role in the control of DNA replication and cell proliferation. The linker histone H1 family members are a key component of chromatin and bind to the nucleosomal core particle around the DNA entry and exit sites. Histone H1 is essential for the condensation of nucleosome chains into higher-order structures. In metazoans, H1 histones are smaller in size with about 200 amino acids. They have a short N-terminal tail, a central globular domain, and a long basic C-terminal tail. The central globular domain is shown to be highly conserved among all H1 subtypes. Manipulating levels of H1 histone can lead to both up- and down-regulation of specific genes. Depletion of H1 results in a massively altered chromosome structure with loss of chromosome banding. In Drosophila, a 20% reduction in H1 content leads to cessation of growth beyond the larval stage. Histone H1 undergoes phosphorylation mainly in the tail region, especially the C-terminal tail, where several {(S/T)-P-X-(K/R)} motifs are located that are recognized by cyclin-dependent kinases (CDKs). Phosphorylation levels are reported to be lowest during the G1 phase of the cell cycle and increase during S phase and reach maximum levels in mitosis. (Ref.: Hergeth, SP., and Schneider, R (2015). EMBO Rep. 16(11); 1439-1453).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H1
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1G6
Conjugate Unconjugated
Dilution ChIP-seq Analysis: A representative lot detected Histone H1 in ChIP-seq applications (Iwasaki, Y.W., et. al. (2016). Mol Cell. 63(3):408-19). Immunoprecipitation Analysis: A representative lot immunoprecipitated Histone H1 in Immunoprecipitation applications (Iwasaki, Y.W., et. al. (2016). Mol Cell. 63(3):408-19).Chromatin Immunoprecipitation Analysis (ChIP): A representative lot immunoprecipitated Histone H1 in Chromatin Immunoprecipitation applications (Iwasaki, Y.W., et. al. (2016). Mol Cell. 63(3):408-19).
Formulation Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias H1
Gene Symbols His 1
Host Species Mouse
Immunogen GST-tagged recombinant fragment corresponding to 148 amino acids from the C-terminal half of Drosophila Histone H1.
Purification Method Protein G purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_001027286
Test Specificity Clone 1G6 is a mouse monoclonal antibody that detects Histone H1 in Drosophila species.
Target Species Drosophila
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.