Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H1T Antibody, Novus Biologicals™
SDP

Catalog No. NBP153001 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP153001 100 μL
NBP1530020 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP153001 Supplier Novus Biologicals Supplier No. NBP153001
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H1T Polyclonal specifically detects Histone H1T in Human samples. It is validated for Western Blot.

Specifications

Antigen Histone H1T
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. P22492
Gene Alias dJ221C16.2, H1 histone family, member T (testis-specific), H1FTMGC163222, H1t, histone 1, H1t, histone cluster 1, H1t, histone H1t, Testicular H1 histone
Gene Symbols HIST1H1T
Host Species Rabbit
Immunogen Synthetic peptides corresponding to HIST1H1T(histone cluster 1, H1t) The peptide sequence was selected from the middle region of HIST1H1T. Peptide sequence KLSKKVIPKSTRSKAKKSVSAKTKKLVLSRDSKSPKTAKTNKRAKKPRAT.
Molecular Weight of Antigen 22 kDa
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 3010
Test Specificity Pig: 86%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Pig, Canine, Rabbit
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.