Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2a, ac Lys5 Antibody (GT1262), Novus Biologicals™
SDP

Catalog No. NB177221 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177221 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177221 Supplier Novus Biologicals Supplier No. NBP313534
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histone H2a Monoclonal antibody specifically detects Histone H2a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen Histone H2a
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone GT1262
Conjugate Unconjugated
Formulation PBS
Gene Alias H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460
Host Species Mouse
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human Histone H2a (acetyl Lys5). The exact sequence is proprietary.
Purification Method Protein G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 8338
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.