Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2a Antibody, Novus Biologicals™
SDP

Catalog No. NB10056346T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056346T 0.025 mg
NB10056346 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056346T Supplier Novus Biologicals Supplier No. NB10056346SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

Histone H2a Polyclonal specifically detects Histone H2a in Human, Mouse, Ovine, Sheep samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot.

Specifications

Antigen Histone H2a
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Flow Cytometry reported in scientific literature (PMID 32849530), ELISA 1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 5 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P04908
Gene Alias H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460
Gene Symbols HIST2H2AC
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides containing amino acid residues 1-15 (MSGRGKQGGKARAKA) and 81-96 (PRHLQLAIRNDEELNK) of human Histone H2A.
Purification Method Protein G purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, Histones and Modified Histones
Primary or Secondary Primary
Gene ID (Entrez) 8338
Test Specificity A 14 kDa band is observed.
Target Species Human, Mouse, Mouse, Ovine, Sheep
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.