Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2A.J Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$156.65 - $620.58

Specifications

Antigen Histone H2A.J
Concentration Lot-Specific
Applications Dot Blot, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3692
SDP
Encompass
View Documents
Active Motif
61794
10 μL
Each for $156.65
Only null left
Add to Cart
 
50-199-3691
SDP
Encompass
View Documents
Active Motif
61793
100 μL
Each for $620.58
Only null left
Add to Cart
 
Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures.
Specifications

Specifications

Histone H2A.J
Dot Blot, Western Blot
Unconjugated
Rabbit
Human, Mouse
Q8R1M2, Q9BTM1
232440, 55766
This antibody was raised against a peptide comprising amino acids 122-129 of human Histone H2A.J.
Primary
-20°C, Avoid Freeze/Thaw Cycles
H2AFJ
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
buforin I; H2A histone family member J; H2A histone family, member J; H2a/j; H2AFJ; H2AJ; Histone H2A.J
H2AFJ
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.