Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H2A.Z Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$156.65 - $638.65
Specifications
Antigen | Histone H2A.Z |
---|---|
Concentration | Conc. Not Determined |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3886
![]() ![]() |
Active Motif
39114 |
10 μL |
Each for $156.65
|
|
|||||
50-227-7220
![]() ![]() |
Active Motif
39013 |
50 μL |
Each for $367.53
|
|
|||||
50-199-3885
![]() ![]() |
Active Motif
39113 |
100 μL |
Each for $638.65
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene encodes a member of the histone H2A family, and generates two transcripts through the use of the conserved stem-loop termination motif, and the polyA addition motif.Specifications
Histone H2A.Z | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
11ac; BOS_6451; C530002L11Rik; H2A histone family member Z; H2A histone family member Z L homeolog; H2A histone family, member V; H2A histone family, member Z; H2A.F/Z; H2A.Z; H2A.Z1; H2A.Z-1; H2a.z-2; H2A.ZK5; H2A.ZK5ac; H2A.Zl; h2a.zl1; h2a.zl2; H2A/z; H2afv; H2afz; h2afz.L; H2AFZac; H2av; H2AZ; H2AZ histone; H2AZK11ac; H2AZK7ac; histone H2A.F/Z; Histone H2A.V; Histone H2A.Z; histone H2A.Zl2; Histone H2A.Z-like; XELAEV_18005732mg; XH2AZ | |
H2AZ1 | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles | |
H2AZ1 |
Conc. Not Determined | |
Unconjugated | |
Rabbit | |
Human | |
P0C0S5 | |
3015 | |
This Histone H2A.Z antibody was raised against a peptide derived from the C-terminus of human histone H2A.Z. | |
Primary | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title