Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AE, p Ser1 Antibody (RM216) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB301032 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301032 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301032 Supplier Novus Biologicals Supplier No. NBP326017
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H2AE Monoclonal antibody specifically detects Histone H2AE in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay

Specifications

Antigen Histone H2AE
Applications Western Blot, ELISA, Immunocytochemistry, Multiplex
Classification Monoclonal
Clone RM216
Conjugate Unconjugated
Dilution Western Blot : 0.5 ug/ml-2 ug/ml, ELISA : 0.2 ug/ml - 1 ug/ml, Immunocytochemistry/ Immunofluorescence : 1 ug/ml - 2 ug/ml, Multiplex Immunoassay : 0.1 ug/ml - 1 ug/ml
Formulation PBS
Gene Alias H2A histone family, member M, H2A/m, H2AFA, H2AFMHistone H2A/m, HIST1H2AE, histone 1, H2ab, histone cluster 1, H2ab, histone H2A type 1-B/E, Histone H2A.2, Histone H2A/a
Host Species Rabbit
Immunogen An unmodified peptide corresponding to the N-terminus of Histone H2AE
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 3012
Target Species Human, Vertebrates
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.