Learn More
Description
Specifications
Specifications
| Antigen | Histone H2Av (p Ser137) |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.1 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:400-1:1600, ELISA 1:29000-1:30000, Immunohistochemistry 2 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin |
| Formulation | 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide |
| Gene Accession No. | P08985 |
| Gene Alias | CG5499 gene product from transcript CG5499-RA, gamma-His2Av, H2AX, 5499, CG5499, CG5499-PA, Dmel\CG5499, gamma-H2Av, gamma-HIS2AV, H2A.F/Z, H2a.V, H2A.Z, H2av, H2AV_DROM, h2AvD, H2AZ, his, His2, HIS2AV, His2AvD, HisH2Av, Hist, Hist2av, Histone H2A variant, l(3)05146, l(3)810, l(3)97Dd, l(3)L1602 |
| Gene Symbols | His2Av |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
