Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB100638V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB100638V 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB100638V Supplier Novus Biologicals Supplier No. NB100638V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Mouse samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot.

Specifications

Antigen Histone H2AX
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias H2A.X, H2A/X, H2AFX
Gene Symbols H2AFX
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to the C-terminus of human histone H2AX using the numbering given for Swiss-Prot entry P16104 (GeneID 3014).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.