Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX Antibody, Novus Biologicals™
SDP

Catalog No. NBP227427SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP227427SS 0.025 mg
NBP227427 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP227427SS Supplier Novus Biologicals Supplier No. NBP227427SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Histone H2AX Polyclonal specifically detects Histone H2AX in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Histone H2AX
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2-4ug/ml∼, Immunohistochemistry-Paraffin 10ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. P16104
Gene Alias H2A.X, H2A/X, H2AFX
Gene Symbols H2AFX
Host Species Rabbit
Immunogen A portion of amino acids 100-143 of human Histone H2A.x was used as the immunogen for the antibody.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Test Specificity The amino acid sequence used as immunogen for the Histone H2A.x antibody is 100% homologous in human and monkey, 94% homologous in cow and dog, 88% homologous in rat and 82% homologous in mouse.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.