Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX, p Ser139 Antibody (2207D), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB349096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB349096 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB349096 Supplier Novus Biologicals Supplier No. FAB2288MFV500
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H2AX Monoclonal specifically detects Histone H2AX in Human samples. It is validated for CyTOF, Flow Cytometry (Intracellular Staining), Western Blot.

Specifications

Antigen Histone H2AX
Applications CyTOF, Flow Cytometry (Intracellular Staining), Western Blot
Classification Monoclonal
Clone 2207D
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias H2A.X, H2A/X, H2AFX
Host Species Rabbit
Immunogen Phospho-peptide containing human Histone H2AX S139 site, Accession # P16104
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.