Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX, p Ser139 Antibody (9F3), Novus Biologicals™
SDP

Catalog No. NBP11925502 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP11925502 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP11925502 Supplier Novus Biologicals Supplier No. NBP1192550.2MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 2 publications

gamma H2AX Monoclonal specifically detects gamma H2AX in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunocytochemistry.

Specifications

Antigen gamma H2AX (p Ser139)
Applications Western Blot
Classification Monoclonal
Clone 9F3
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
Gene Accession No. P16104
Gene Alias H2A histone family, member X, H2A.X, H2a/x, H2AX histone, H2AXhistone H2A.x, YH2AX
Gene Symbols H2AFX
Host Species Mouse
Immunogen Synthetic phospho-peptide derived from the sequence of human Histone H2AX phosphorylated at Ser139.
Molecular Weight of Antigen 15 kDa
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline DNA Repair, Mitotic Regulators, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Guinea Pig, Hamster, Primate, Rabbit, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.