Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AX, p Ser139 Antibody (BLR053F), Novus Biologicals™
SDP

Catalog No. NB188018 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB188018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB188018 Supplier Novus Biologicals Supplier No. NBP314678
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)

Specifications

Antigen Histone H2AX
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone BLR053F
Conjugate Unconjugated
Dilution Western Blot 1:1000, Flow Cytometry, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:500, Immunoprecipitation 20 ul/1mg lysate, Immunohistochemistry-Paraffin 1:100 - 1:500
Formulation Borate Buffered Saline (BBS) pH 8.2 with 0.1% BSA and 0.09% Sodium Azide
Gene Alias H2A.X, H2A/X, H2AFX
Host Species Rabbit
Immunogen surrounding Serine 140
Quantity 100 μg
Regulatory Status RUO
Research Discipline Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 3014
Target Species Human, Mouse
Content And Storage Store at 2 - 8°C/ 1 year from date of receipt
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.