Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2AY/macroH2A.1 Antibody (RM248), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB294145 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB294145 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB294145 Supplier Novus Biologicals Supplier No. NBP326031IR
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Histone H2AY/macroH2A.1 Monoclonal antibody specifically detects Histone H2AY/macroH2A.1 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay

Specifications

Antigen Histone H2AY/macroH2A.1
Applications Western Blot, ELISA, Immunocytochemistry, Multiplex
Classification Monoclonal
Clone RM248
Conjugate DyLight 755
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
Formulation 50mM Sodium Borate
Gene Alias core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1
Host Species Rabbit
Immunogen A peptide corresponding to the C-terminus of Human Histone H2AY/macroH2A.1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9555
Target Species Human, Vertebrates
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.