Learn More
Description
Specifications
Specifications
| Antigen | Histone H2B |
| Applications | Western Blot, ELISA, Dot Blot, Immunofluorescence, ChIP Assay |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:1000, ELISA 1:1000, Dot Blot 1:5000, Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 0.5-1 ug, Chromatin Immunoprecipitation Sequencing |
| Formulation | PBS with 0.05% Sodium Azide and 0.05% ProClin 300 |
| Gene Alias | FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 |
| Gene Symbols | HIST2H2BF |
| Host Species | Rabbit |
| Immunogen | The exact immunogen is propietary information. |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
