Learn More
Histone H2B, member S Antibody, Novus Biologicals™
SDP
Rabbit Polyclonal Antibody has been used in 13 publications
Supplier:  Novus Biologicals NB10056347SS
 View more versions of this product
Catalog No. NB10056347T
Only null left
Add to Cart
Description
Description
Histone H2B, member S Polyclonal specifically detects Histone H2B, member S in Human, Mouse, Rat, Canine, Chicken, Drosophila, Mammal, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
Specifications
Specifications
Histone H2B, member S
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
H2BFS
This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acid residues 111-125 AVSEGTKAVTKYTSS of human Histone H2B; GenBank no. CAI19747.1.
0.025 mg
Chromatin Research
54145
Human, Mouse, Rat, Canine, Chicken, Drosophila, Mammalia, Zebrafish
Purified
Western Blot, Immunohistochemistry (Paraffin), Immunoblot
1.0 mg/mL
Western Blot 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml, Immunoblotting reported in scientific literature (PMID 28235947)
H2B histone family S member10H2B/s, H2B histone family, member S, Histone H2B.s
Rabbit
Protein G purified
RUO
Primary
A 15 kDa band is observed. This antibody is routinely quality controlled in human lysates.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only