Learn More
Histone H2B, member S, p Ser14 Antibody (C.15200186), Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | HistoneH2B,memberS |
| Applications | Dot Blot, ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | C.15200186 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:200, ELISA 1:100, Dot Blot 1:2000 |
| Formulation | PBS with 0.05% Sodium Azide and 0.05% ProClin 300 |
| Gene Alias | H2B histone family S member10H2B/s, H2B histone family, member S, Histone H2B.s |
| Gene Symbols | H2BFS |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.