Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H2B Rabbit anti-Human, Mouse, Rat, Clone: 3U2P5, Novus Biologicals™
SDP

Catalog No. NB166292 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166292 20 μg
NB166291 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166292 Supplier Novus Biologicals Supplier No. NBP31559820UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H2B Monoclonal antibody specifically detects Histone H2B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen Histone H2B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 3U2P5
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 47-126 of human Histone H2B (O60814).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics
Primary or Secondary Primary
Gene ID (Entrez) 440689
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.